Learn More
Sino Biological Recombinant Human Prostasin/PRSS8 Protein (His Tag)
Supplier:  Sino Biological 10287H08H50
Encompass_Preferred
A DNA sequence encoding the mature form of human Prss8 (NP_002764.1) (Ala 30-Arg 322) was fused with a signal peptide at the N-terminus and a polyhistidine-tag at the C-terminus.
Catalog No. 50-224-6953