Learn More
American Research Products Inc Human prostate specific antige
Catalog No. 5000326565
Encompass
Human prostate specific antigen PSA ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26565 Supplier American Research Products Inc Supplier No. CSBE04768H
Human prostate specific antigen PSA ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich