Learn More
R&D Systems™ Human Prostatic Acid Phosphatase/ACPP DuoSet ELISA

Description
Specifications
Specifications
| Assay Range | 62.50 to 4000 pg/mL |
| Product Type | Solid Phase Sandwich ELISA |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 55 |
| Gene Symbol | ACPP |
| Host Species | Human |
| Quantity | 1 Kit (for 5 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | acid phosphatase, prostate, ACP3, ACP-3, ACPP, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase |
| Target | Prostatic Acid Phosphatase/ACPP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.