Learn More
Biomatik Corporation Human Protease Activated Receptor 2 (PAR2) CLIA Kit
Supplier:  Biomatik Corporation EKU09894
Encompass_Preferred
Human Protease Activated Receptor 2 (PAR2) CLIA Kit for detecting PAR2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P55085. Gene ID: 2150. Target Synonyms: F2RL1; F2-RL1; GPR11; PAR2; Coagulation Factor II (thrombin) receptor-Like 1; G-protein coupled receptor 11; Thrombin receptor-like 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8406