Learn More
Biomatik Corporation Human Protease, Serine 23 (PRSS23) ELISA Kit
Supplier:  Biomatik Corporation EKU06879
Encompass_Preferred
Human Protease, Serine 23 (PRSS23) ELISA Kit for detecting PRSS23 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95084. Gene ID: 11098. Target Synonyms: SPUVE; SIG13; ZSIG13; Putative secreted protein Zsig13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8802