Learn More
Biomatik Corporation Human Proteasome 26S Subunit, Non ATPase 10 (PSMD10) ELISA Kit
Supplier:  Biomatik Corporation EKU06883
Encompass_Preferred
Human Proteasome 26S Subunit, Non ATPase 10 (PSMD10) ELISA Kit for detecting PSMD10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75832. Gene ID: 5716. Target Synonyms: p28; Gankyrin; 26S proteasome non-ATPase regulatory subunit 10; 26S proteasome regulatory subunit p28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6971