Learn More
Biomatik Corporation Human Proteasome Activator Subunit 3 (PSME3) ELISA Kit
Supplier:  Biomatik Corporation EKU06884
Encompass_Preferred
Human Proteasome Activator Subunit 3 (PSME3) ELISA Kit for detecting PSME3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P61289. Gene ID: 10197. Target Synonyms: Ki; PA28G; REG-GAMMA; 11S regulator complex subunit gamma; Activator of multicatalytic protease subunit 3; Ki nuclear autoantigen; Proteasome activator 28 subunit gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4597