Learn More
Biomatik Corporation Human Proteasome Subunit Alpha Type 6 (PSMa6) ELISA Kit
Supplier:  Biomatik Corporation EKU12054
Encompass_Preferred
Human Proteasome Subunit Alpha Type 6 (PSMa6) ELISA Kit for detecting PSMa6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60900. Gene ID: 5687. Target Synonyms: IOTA; PROS27; p27K; 27 kDa prosomal protein; Macropain iota chain; Proteasome iota chain; Multicatalytic endopeptidase complex iota chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7248