Learn More
Biomatik Corporation Human Proteasome Subunit Alpha Type 5 (PSMa5) ELISA Kit
Supplier:  Biomatik Corporation EKU06886
Encompass_Preferred
Human Proteasome Subunit Alpha Type 5 (PSMa5) ELISA Kit for detecting PSMa5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28066. Gene ID: 5686. Target Synonyms: PSC5; Macropain zeta chain; Multicatalytic endopeptidase complex zeta chain; Proteasome zeta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5630