Learn More
American Research Products Inc Human protein arginine deimina
Catalog No. 5000326657
Encompass
Human protein-arginine deiminase type-4 (PADI4) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum urine cell culture supernates Method Sandwich
Catalog No. 50-003-26657 Supplier American Research Products Inc Supplier No. CSBE16219H
Human protein-arginine deiminase type-4 (PADI4) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum urine cell culture supernates Method Sandwich