Learn More
Biomatik Corporation Human Protein C Inhibitor (PCI) CLIA Kit
Supplier:  Biomatik Corporation EKU09857
Encompass_Preferred
Human Protein C Inhibitor (PCI) CLIA Kit for detecting PCI in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 109.7-80,000pg/ml. Sensitivity: 41.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05154. Gene ID: 5104. Target Synonyms: SERPINA5; PAI3; PLANH3; PROCI; Serpin Peptidase Inhibitor Clade A 5; Plasma Serine Protease Inhibitor; Plasminogen Activator Inhibitor 3; Acrosomal serine protease inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5610