Learn More
Biomatik Corporation Human Protein Disulfide Isomerase A5 (PDIA5) ELISA Kit
Supplier:  Biomatik Corporation EKU06905
Encompass_Preferred
Human Protein Disulfide Isomerase A5 (PDIA5) ELISA Kit for detecting PDIA5 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14554. Gene ID: 10954. Target Synonyms: PDI-A5; PDIR; Protein Disulfide Isomerase-Associated 5; Protein disulfide isomerase-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5634