Learn More
Biomatik Corporation Human Protein Disulfide Isomerase (PDI) ELISA Kit
Supplier:  Biomatik Corporation EKU06900
Encompass_Preferred
Human Protein Disulfide Isomerase (PDI) ELISA Kit for detecting PDI in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13087. Gene ID: 64714. Target Synonyms: PDI-A2; PDA2, PDIA2, PDIR; PDIP; Protein Disulfide Isomerase,Pancreatic; Protein Disulfide Isomerase-Associated 2; Pancreas-specific protein disulfide isomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7629