Learn More
Biomatik Corporation Human Protein Kinase, AMP Activated Gamma 2 (PRKAg2) ELISA Kit
Supplier:  Biomatik Corporation EKU12356
Encompass_Preferred
Human Protein Kinase, AMP Activated Gamma 2 (PRKAg2) ELISA Kit for detecting PRKAg2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UGJ0. Gene ID: 51422. Target Synonyms: AAKG; AAKG2; CMH6; H91620p; WPWS; 5'-AMP-activated protein kinase subunit gamma-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7978