Learn More
Bioworld HUMAN PROTEIN KINASE C-E (R
Supplier:  Bioworld 220604621
Encompass_Preferred
HUMAN PROTEIN KINASE C-E (RECOMBINANT): Recombinant Human Protein Kinase C epsilon (PKC-e) Source: SF9 insect cells. Description : Recombinant Human PKC-epsilon produced in Sf9 is a glycosylated, polypeptide chain containing amino ac...
Catalog No. 50-253-559