Learn More
Biomatik Corporation Human Protein Kinase C Epsilon (PKCe) CLIA Kit
Supplier:  Biomatik Corporation EKU09915
Encompass_Preferred
Human Protein Kinase C Epsilon (PKCe) CLIA Kit for detecting PKCe in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.14-100ng/ml. Sensitivity: 0.07ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q02156. Gene ID: 5581. Target Synonyms: PKC-E; PRKCE; PRKC-E; NPKC-Epsilon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4932