Learn More
Biomatik Corporation Human Protein Phosphatase 2, Catalytic Subunit Alpha Isoform (PPP2Ca) ELISA Kit
Supplier:  Biomatik Corporation EKU12030
Encompass_Preferred
Human Protein Phosphatase 2, Catalytic Subunit Alpha Isoform (PPP2Ca) ELISA Kit for detecting PPP2Ca in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P67775. Gene ID: 5515. Target Synonyms: PP2CA; PP2Ac; RP-C; Replication protein C; Serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7292