Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Protein S Native Protein

Catalog No. PIRP43131 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43131 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43131 Supplier Invitrogen™ Supplier No. RP43131

missing translation for 'validMainframeMsg'

Native Protein

Protein S (human) Native Protein, RP-43131, from Invitrogen.

RP-43131 contains native human Protein S protein. RP-43131 may be used as a positive control in many immunological assays. The RP-43131 protein molecular weight is ∽69 kDa. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07225
Concentration 6.1mg/mL
For Use With (Application) Control
Formulation 50%water with 50% glycerol and no preservative
Gene ID (Entrez) 5627
Molecular Weight (g/mol) ∽69
Name Human Protein S
Quantity 100 μg
Source Human
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name Human Protein S
Gene Symbol PROS1
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species Human
Recombinant Native
Form Liquid
Protein Subtype Native
Purity or Quality Grade purified
Show More Show Less

in vitro research only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.