Learn More
Biomatik Corporation Human Protein SCAF11 (SCAF11) ELISA Kit
Supplier:  Biomatik Corporation EKU10083
Encompass_Preferred
Human Protein SCAF11 (SCAF11) ELISA Kit for detecting SCAF11 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99590. Gene ID: 9169. Target Synonyms: SRSF2IP; SFRS2IP; SIP1; SRRP129; Cysteinyl Aspartate Specific Proteinases 11; Apoptosis-Related Cysteine Peptidase 11; Splicing Factor,Arginine/Serine-Rich 2,Interacting. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7217