Learn More
Biomatik Corporation Human Protein Tyrosine Phosphatase Like Protein A (PTPLA) ELISA Kit
Supplier:  Biomatik Corporation EKU06944
Encompass_Preferred
Human Protein Tyrosine Phosphatase Like Protein A (PTPLA) ELISA Kit for detecting PTPLA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B0YJ81. Gene ID: 9200. Target Synonyms: CAP; HACD1; Proline Instead Of Catalytic Arginine Member a; 3-hydroxyacyl-CoA dehydratase 1; Cementum Attachment Protein; Very-long-chain 3R-3-hydroxyacyl-CoA dehydratase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8794