Learn More
Biomatik Corporation Human Protein Tyrosine Phosphatase Receptor Type G (PTPRG) ELISA Kit
Supplier:  Biomatik Corporation EKU06945
Encompass_Preferred
Human Protein Tyrosine Phosphatase Receptor Type G (PTPRG) ELISA Kit for detecting PTPRG in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23470. Gene ID: 5793. Target Synonyms: PTPR-G; HPTPG; PTPG; R-PTP-GAMMA; RPTPG; Receptor-type tyrosine-protein phosphatase gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8793