Learn More
Biomatik Corporation Human Protein Tyrosine Phosphatase, Non Receptor Type 22 (PTPN22) ELISA Kit
Supplier:  Biomatik Corporation EKU12442
Encompass_Preferred
Human Protein Tyrosine Phosphatase, Non Receptor Type 22 (PTPN22) ELISA Kit for detecting PTPN22 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2R2. Gene ID: 26191. Target Synonyms: LYP; PEP; Lyp1; Lyp2; PTPN8; Lymphoid phosphatase; Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP; PEST-domain phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6149