Learn More
Biomatik Corporation Human Protein Z (PROZ) ELISA Kit
Supplier:  Biomatik Corporation EKU06952
Encompass_Preferred
Human Protein Z (PROZ) ELISA Kit for detecting PROZ in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22891. Gene ID: 8858. Target Synonyms: PRO-Z; PZ; Vitamin K Dependent Protein Z; VKDPZ; VKDP-Z. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4859