Learn More
Bon Opus Biosciences Recombinant Human Prothrombin (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP106350UG
Encompass_Preferred
Recombinant Human Prothrombin (C-hFc) is produced by Human Cells. The target gene encoding Ala1-Glu579 is expressed with a C-hFc tag. Accession# AAH51332. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1063
Catalog No. 50-217-2661