Learn More
American Research Products Inc Human Proto oncogene Wnt 3 WN
Catalog No. 5000327353
Encompass
Human Proto-oncogene Wnt-3 (WNT3) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27353 Supplier American Research Products Inc Supplier No. CSBEL026135HU
Human Proto-oncogene Wnt-3 (WNT3) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich