Learn More
Raybiotech Inc Human Protocadherin-17 ELISA Kit
Supplier:  Raybiotech Inc ELHPCDH171
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.045 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Protocadherin-17.
Catalog No. 50-194-9624