Learn More
Biomatik Corporation Human Protocadherin 20 (PCDH20) ELISA Kit
Supplier:  Biomatik Corporation EKU08319
Encompass_Preferred
Human Protocadherin 20 (PCDH20) ELISA Kit for detecting PCDH20 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.126ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N6Y1. Gene ID: 64881. Target Synonyms: PCDH13; Protocadherin-13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8680