Learn More
Biomatik Corporation Human Protocadherin Gamma A2 (PCDHgA2) ELISA Kit
Supplier:  Biomatik Corporation EKU06972
Encompass_Preferred
Human Protocadherin Gamma A2 (PCDHgA2) ELISA Kit for detecting PCDHgA2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5H1. Gene ID: 56113. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8792