Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PRP19 (aa 176-270) Control Fragment Recombinant Protein

Catalog No. RP103345
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP103345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP103345 Supplier Invitrogen™ Supplier No. RP103345
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84400. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PSO4 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UMS4
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 27339
Name Human PRP19 (aa 176-270) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AA617263; AL024362; D19Wsu55e; hPSO4; Neuronal differentiation-related gene protein; NMP200; Nuclear matrix protein 200; nuclear matrix protein NMP200 related to splicing factor PRP19; nuclear matrix protein SNEV; pre-mRNA processing factor 19; pre-mRNA-processing factor 19; PRP19; prp19 beta; PRP19/PSO4 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae); PRPF19; PSO4; psoralen 4; RING-type E3 ubiquitin transferase PRP19; Senescence evasion factor; SNEV; UBOX4
Common Name PRP19
Gene Symbol Prpf19
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LQDKATVLTTERKKRGKTVPEELVKPEELSKYRQVASHVGLHSASIPGILALDLCPSDTNKILTGGADKNVVVFDKSSEQILATLKGHTKKVTSV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.