Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PRPS2 Control Fragment Recombinant Protein

Catalog No. RP105432
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP105432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP105432 Supplier Invitrogen™ Supplier No. RP105432
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66936. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The Phosphoribosyl Pyrophosphate Synthetase 2 (PRPS2) gene plays a critical role in cellular metabolism by encoding an enzyme that conducts a key reaction linking central carbon metabolism and nucleotide production, essential for various biological functions. Structurally, PRPS2 shares a high sequence identity with PRPS1, making it less sensitive to allosteric inhibition and particularly significant in certain oncology contexts due to its upregulation at the translational level in some cancers. The enzyme encoded by PRPS2 is pivotal in coupling protein and nucleotide biosynthesis, aided by a unique cis-regulatory element in its 5' UTR, which is influenced by oncogenic pathways. This gene manifests a tissue-specific expression profile, predominantly in proliferative tissues such as the thymus, lung, and small intestine, highlighting its significance in cell growth and proliferation. The evolutionary origins of PRPS2, along with its functional diversification into multiple PRPS genes in eukaryotes, underscore its indispensable role in cellular physiology and pathology.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11908
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 5634
Name Human PRPS2 Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2610101M19Rik; AA589463; AI464149; Phophoribosylpyrophosphate synthetase, subunit II; phosphoribosyl pyrophosphate synthase II; phosphoribosyl pyrophosphate synthetase 2; phosphoribosyl pyrophosphate synthetase II; PPRibP; PPRibP synthetase; Prps2; Prps-2; PRSII; PRS-II; ribose-phosphate diphosphokinase 2; ribose-phosphate pyrophosphokinase 2; testicular secretory protein Li 41
Common Name PRPS2
Gene Symbol PRPS2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GAKRYHAGYTLQISPSADWF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.