Learn More
Sino Biological Anti-PRSS2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10813RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Trypsin 2 / PRSS2 (rh Trypsin 2 / PRSS2; Catalog#10813-H08H; NP_002761.1; Met1-Ser247). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-7654