Learn More
Abcam Human PSA ELISA Kit, 1 x 384 Tests
Catalog No. 502656689
Encompass_Preferred
Human PSA ELISA Kit is a single-wash 90-min Simplestep used to quantify Human PSA with a sensitivity of 8 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6689 Supplier Abcam Supplier No. AB2646151X384TESTS
Human PSA ELISA Kit is a single-wash 90-min Simplestep used to quantify Human PSA with a sensitivity of 8 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement