Learn More
BIOHIPPO Human PSA Microsample
SDP
Catalog No. 5000313763
Encompass
Human PSA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PSA in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 096 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PSA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PSA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PSA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13763 Supplier BIOHIPPO Supplier No. BHE15210481
Human PSA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PSA in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 096 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PSA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PSA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PSA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color