Learn More
Sino Biological Recombinant Human PSAP/Prosaposin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 16224H08H1
Encompass_Preferred
A DNA sequence encoding the human PSAP (NP_002769.1) (Met1-Asn524) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-8131