Learn More
Sino Biological Human PSGL-1/CD162 HEK293 Overexpression Lysate
Catalog No. NC3852463
Encompass_Preferred
This Human PSGL-1/CD162 overexpression lysate was created in HEK293 Cells and intented for use as a Western blot (WB) positive control. Purification of PSGL-1/CD162 protein (Cat: 10490-H05H) from the overexpression lysate was verified.
Catalog No. NC3852463 Supplier Sino Biological Supplier No. 10490H05HL300/DEL
This Human PSGL-1/CD162 overexpression lysate was created in HEK293 Cells and intented for use as a Western blot (WB) positive control. Purification of PSGL-1/CD162 protein (Cat: 10490-H05H) from the overexpression lysate was verified.