Learn More
Sino Biological Recombinant Human OX40/TNFRSF4 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10481H02H1
Encompass_Preferred
A DNA sequence encoding the human TNFRSF4 (NP_003318.1) (Met1-Ala216) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-222-3543