Learn More
ACROBiosystems Human PSMA 200ug
Supplier:  ACROBiosystems PSAH82F7200UG
Encompass_Preferred
Biotinylated Human PSMA Protein FcAvitag (PSA-H82F7) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1).
Catalog No. 50-253-4280