Learn More
ACROBiosystems Human PSMA 25ug
Catalog No. 502534279
Encompass_Preferred
Biotinylated Human PSMA Protein FcAvitag (PSA-H82F7) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1).
Catalog No. 50-253-4279 Supplier ACROBiosystems Supplier No. PSAH82F725UG
Biotinylated Human PSMA Protein FcAvitag (PSA-H82F7) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1).