Learn More
Sino Biological Anti-PSMD5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 108177T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PSMD5, and purified by antigen affinity chromatography.
Catalog No. 50-240-8857