Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PAF ELISA Kit

Catalog No. EEL197
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL197 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL197 Supplier Invitrogen™ Supplier No. EEL197
Add to Cart
Add to Cart

ELISA

Human PAF ELISA Kit quantifies PAF in serum and plasma. The Human PAF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Platelet Activating Factor (PAF) is a potent phospholipid mediator involved in various biological processes, including inflammation, thrombosis, and allergic reactions. PAF is synthesized by various cell types, including platelets, endothelial cells, and leukocytes. It exerts its effects by binding to the PAF receptor (PAFR), a G-protein coupled receptor, triggering a cascade of intracellular signaling pathways.PAF plays a critical role in the regulation of platelet aggregation, vascular permeability, and leukocyte activation. It is implicated in numerous pathological conditions, such as asthma, sepsis, cardiovascular diseases, and cancer. Due to its significant role in inflammation and immune responses, PAF is considered a valuable target for therapeutic interventions aimed at modulating its activity to treat associated diseases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P25105
Assay Range 78.125-5000 pg/mL
Assay Sensitivity 46.875 pg/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Azelaoyl-PAF; C33H66NO9PAzelaoyl-PAF; C33H66NO9P;
Gene ID (Entrez) 5724
Gene Symbol PTAFR
Interassay CV <10%
Intraassay CV <10%
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard , Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis, Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Shipping Condition Wet Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target PAF
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.