Learn More
Biomatik Corporation Human PTEN Induced Putative Kinase 1 (PINK1) ELISA Kit
Supplier:  Biomatik Corporation EKU12084
Encompass_Preferred
Human PTEN Induced Putative Kinase 1 (PINK1) ELISA Kit for detecting PINK1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BXM7. Gene ID: 65018. Target Synonyms: BRPK; PARK6; Parkinson Disease (Autosomal Recessive) 6; Serine/threonine-protein kinase PINK1, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8034