Learn More
BIOHIPPO Human PTGFR Microsample
SDP
Catalog No. 5000313998
Encompass
Human PTGFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PTGFR in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 2 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PTGFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PTGFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PTGFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13998 Supplier BIOHIPPO Supplier No. BHE15212411
Human PTGFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PTGFR in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 78-50 ng/mL with a sensitivity of 0 2 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PTGFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PTGFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PTGFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color