Learn More
Sino Biological Human PTH PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13192T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Parathyroid Hormone/PTH (rh Parathyroid Hormone/PTH; Catalog#13192-H09E; P01270; Ser32-Phe65). Parathyroid Hormone/PTH specific IgG was purified by Human Parathyroid Hormone/PTH affinity chromatography.
Catalog No. 50-237-3653