Learn More
Sino Biological Recombinant Human Parathyroid Hormone/PTH Protein (hFc Tag)
Supplier:  Sino Biological 13192H01H1
Encompass_Preferred
A DNA sequence encoding the human PTH (P01270) (Ser32-Gln115) was expressed,with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-255-4291