Learn More
Raybiotech Inc Human PTH1R ELISA Kit
Supplier:  Raybiotech Inc ELHPTH1R1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 8.2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human PTH1R.
Catalog No. 50-194-9670