Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human PTH1R ELISA Kit

Catalog No. EH385RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH385RBX5 5 x 96 Tests
EH385RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH385RBX5 Supplier Invitrogen™ Supplier No. EH385RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human PTH1R quantitates human PTH1R in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PTH1R.

Parathyroid hormone receptor 1 is a member of the G-protein coupled receptor family. This protein is a receptor for parathyroid hormone (PTH) and for parathyroid hormone-like hormone (PTHLH). The activity of this receptor is mediated by G proteins which activate adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system. Defects in this receptor are known to be the cause of Jansen's metaphyseal chondrodysplasia (JMC), chondrodysplasia Blomstrand type (BOCD), as well as enchodromatosis. Parathyroid hormone receptor 1 has been reported to be expressed in bone, breast, brain, colon, GI, kidney, liver, placenta, skin, umbilical cord, and uterus.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q03431
Assay Range 8.19-2000 pg/mL
Assay Sensitivity 8.2 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias MGC138426; MGC138452; Parathormone; Parathyrin; parathyroid hormone 1 receptor; parathyroid hormone receptor; parathyroid hormone receptor 1; parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PFE; PPR; PTH; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTH1R; Pthr; Pthr1; PTHrel; PTH-related peptide receptor; PTHrP-R; PTHrP-receptor; seven transmembrane helix receptorMGC138426; MGC138452; Parathormone; Parathyrin; parathyroid hormone 1 receptor; parathyroid hormone receptor; parathyroid hormone receptor 1; parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PFE; PPR; PTH; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTH1R; Pthr; Pthr1; PTHrel; PTH-related peptide receptor; PTHrP-R; PTHrP-receptor; seven transmembrane helix receptor;
Gene ID (Entrez) 5745
Gene Symbol Pth1r
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target PTH1R
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.