Learn More
ACROBiosystems ACROBiosystems Human PTH1R / PTHR1 Protein, His Tag
Catalog No. 502106749
Encompass_Preferred
Recombinant Protein;100UG;Human PTH1R, His Tag (PTR-H5222) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Gly 188 (Accession # Q03431-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.6 kDa. The protein migrates as 27-43 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-210-6749 Supplier ACROBiosystems Supplier No. PTRH5222100UG
Recombinant Protein;100UG;Human PTH1R, His Tag (PTR-H5222) is expressed from human 293 cells (HEK293). It contains AA Asp 27 - Gly 188 (Accession # Q03431-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.6 kDa. The protein migrates as 27-43 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.