Learn More
Sino Biological Anti-Ptk2b Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106029T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Ptk2b, and purified by antigen affinity chromatography.
Catalog No. 50-240-9719