Learn More
ACROBiosystems ACROBiosystems Human PTK7 / CCK4 Protein, Fc Tag
Supplier:  ACROBiosystems PT7H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human PTK7, Fc Tag (PT7-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 31 - Thr 704 (Accession # Q13308-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101.1 kDa.
Catalog No. 50-210-6378