Learn More
ACROBiosystems Human PTK7 / CCK4 Protein, His Tag
Catalog No. 502054798
Encompass_Preferred
Recombinant Protein;100UG;Human PTK7, His Tag (PT7-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 31 - Thr 704 (Accession # Q13308-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.5 kDa.
Catalog No. 50-205-4798 Supplier ACROBiosystems Supplier No. PT7H52H3100UG
Recombinant Protein;100UG;Human PTK7, His Tag (PT7-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 31 - Thr 704 (Accession # Q13308-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.5 kDa.